Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ FetalClone™ II Serum (U.S.)

Catalog No. SH3006602HI Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
HI, Irr - 25 to 40 doses
Heat Inactivated
Irradiated (25 to 40 doses)
Irradiated (30 to 45 doses)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3006602HI 100 mL Heat Inactivated
SH366IH2540 500 mL HI, Irr - 25 to 40 doses
SH300660340 500 mL Irradiated (25 to 40 doses)
SH3663IR345 500 mL Irradiated (30 to 45 doses)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SH3006602HI Supplier Cytiva Supplier No. SH30066.02HI
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Optimized for the growth of CHO cells and derivatives.

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
  • demonstrated performance with a variety of cell lines, including hybridomas, CHO, BHK-21, NS0, MRC-5, and Vero cells
  • TRUSTED_SUSTAINABILITY

    Specifications

    Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
    Country of Origin United States
    Product Type FBS Replacement Serum
    Quantity 100 mL
    Sterility Sterile
    Sterilization Method Sequential-filtered, 100 nm
    Processing Heat Inactivated
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 10 EU/mL
    Heat Inactivated Yes
    Hemoglobin Concentration ≤ 20 mg/dL
    IgG levels Typically less than 2 μg/mL
    Product Line HyClone
    Mycoplasma Detected No
    USDA Safety Testing Included USDA Inspected
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.