Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Hank's 1X Balanced Salt Solutions

Catalog No. SH3003103 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
6 x 500 mL
6 x 1000 mL
With Additives:
Calcium, Magnesium
Calcium, Magnesium, Phenol Red
Phenol Red
Without Additives:
No Calcium, No Magnesium
No Calcium, No Magnesium, No Phenol Red
No Phenol Red
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives
SH3003103 1000 mL Phenol Red No Calcium, No Magnesium
SH3058802 1000 mL - No Calcium, No Magnesium, No Phenol Red
SH30031FS 6 x 500 mL Phenol Red No Calcium, No Magnesium
SH3026801 500 mL Calcium, Magnesium No Phenol Red
SH3003102 500 mL Phenol Red No Calcium, No Magnesium
SH3058801 500 mL - No Calcium, No Magnesium, No Phenol Red
SH3003101 100 mL Phenol Red No Calcium, No Magnesium
SH3003002 500 mL Calcium, Magnesium, Phenol Red -
SH3026802 1000 mL Calcium, Magnesium No Phenol Red
SH30268LS 6 x 1000 mL Calcium, Magnesium No Phenol Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. SH3003103 Supplier Cytiva Supplier No. SH30031.03
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Buffer cell culture growth with 1X balanced salt solutions

  • Used for washing and treating tissues and cells
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance cGMP and ISO 9001:2000 Guidelines
Content And Storage 15° to 30°C
Concentration 1 X
Form Liquid
Product Type Hanks' Balanced Salt Solution
Sterilization Method 0.1 μm Sterile-filtered
With Additives Phenol Red
Without Additives No Calcium, No Magnesium
Endotoxin Level =1.0EU/mL
pH 7.0 to 7.4
Quantity 1000 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.