Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ HyCell CHO Powder Cell Culture Medium without L-glutamine and with HT

Catalog No. p-7145856 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 L
1000 mL PETE bottle
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3093401 1000 mL PETE bottle
SH3093302 10 L
SH3093301 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3093401 Supplier Cytiva Supplier No. SH30934.01
Only null left
Add to Cart
Add to Cart

A versatile, animal derived component free (ADCF) cell culture media.

  • Chemically defined and animal-derived component‑free formulations
  • Designed for high cell yield and recombinant protein production
  • Allow direct or sequential adaptation
  • Designed for large-scale culture applications, including perfusion and fed-batch strategies
  • Developed for Chinese hamster ovary (CHO) cell culture in bioprocessing applications
  •  Specifically formulated to provide excellent performance, support high peak cell density, and enhance product yield and quality
  • Can be used for batch, fed-batch, or perfusion applications in multiple cell culture platforms ranging from shake flasks to large-scale bioreactors
  • TRUSTED_SUSTAINABILITY

    Specifications

    Form Powder
    Sterility Non-sterile
    With Additives Sodium Bicarbonate, Poloxamer 188, Hypoxanthine, Thymidine
    Without Additives No Phenol Red, No HEPES, No Glutamine
    Quantity 1000 mL PETE bottle
    Content And Storage 2° to 8°C, away from light and moisture
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.