Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Iron-Supplemented Calf Serum, U.S. origin

Catalog No. SH300720340 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Heat Inactivated
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses
None
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH300720340 500 mL Irradiated, 25 to 40 doses
SH300720440 1000 mL Irradiated, 25 to 40 doses
SH3007202 100 mL None
SH300720445 1000 mL Irradiated, 30 to 45 doses
SH3007202HI 100 mL Heat Inactivated
SH3007204HI 1000 mL Heat Inactivated
SH3007203 500 mL None
SH3007204 1000 mL None
SH3007203HI 500 mL Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. SH300720340 Supplier Cytiva Supplier No. SH30072.03IR2540
Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

A cost-effective alternative to Fetal Bovine Serum (FBS). Contains additional iron to boost serum transferrin to physiological levels.

HyClone Iron-Supplemented Bovine Calf Serum, US Origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. This serum is carefully supplemented with iron to load serum transferrin levels to physiological levels. After supplementation, the serum contains three to four times as much available iron and transferrin as fetal bovine serum (FBS) or equine serum.

TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing Irradiated, 25 to 40 doses
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Heat Inactivated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.