Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum (New Zealand)

Catalog No. SH3062603HI Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1000 mL
Processing:
Heat Inactivated
None
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3062603HI 1000 mL Heat Inactivated
SH3062603 1000 mL None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3062603HI Supplier Cytiva Supplier No. SH30626.03HI

missing translation for 'validMainframeMsg'

A cost-effective alternative to Fetal Bovine Serum (FBS).

  • Ensured safety against bovine diseases
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Ensured uniformity and consistency between bottles
  • TRUSTED_SUSTAINABILITY

    Specifications

    Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG
    Buffer Lactate Dehydrogenase
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
    Country of Origin New Zealand
    Additive Alkaline Phosphatase
    Product Type Newborn Calf Bovine Serum
    Quantity 1000 mL
    Sterility Sterile
    Sterilization Method Sequential-filtered, 100 nm
    Processing Heat Inactivated
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 25 EU/mL
    Heat Inactivated Yes
    Hemoglobin Concentration ≤ 25 mg/dL
    Product Line HyClone
    USDA Safety Testing Included Yes
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.