Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ L-Glutamine, Powder

Catalog No. SH3033602 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3033602 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3033602 Supplier Cytiva Supplier No. SH30336.02

missing translation for 'validMainframeMsg'

For supplementing serum- and protein-free media

  • Support the growth of cells that have high energy demands
  • Synthesize large amounts of proteins and nucleic acids
TRUSTED_SUSTAINABILITY

Specifications

Amino Acids L-Glutamine
Form Powder
Product Type L-Glutamine
For Use With (Application) Supplementing Serum- and Protein-Free Media
Product Line HyClone
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.