Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Newborn Bovine Calf Serum, Heat Inactivated

Catalog No. SH3011802HI Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3011802HI 100 mL
SH3011803HI 500 mL
SH3011804HI 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3011802HI Supplier Cytiva Supplier No. SH30118.02HI
Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Augment cell culture growth with this cost-effective alternative to Fetal Bovine Serum (FBS)

  • Produced using careful collection and processing procedures, including venipuncture, to provide the highest quality product available
  • Ensures excellent culture results without fear of contamination
  • HyClone™ maintains complete control of the entire collection and production process for 100% traceability
  • Newborn bovine calf serum undergoes the same careful collection and processing procedures (including venipuncture) used for other bovine serum products
  • Sourced from the United States with complete traceability back to original source
  • Age of animal at time of collection less than 10 days old
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or below -10°C
Age <10 days
Country of Origin United States
Form Liquid
Product Type Newborn Calf Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing Heat Inactivated
Volume (Metric) 100 mL
Endotoxin Concentration ≤ 50 EU/mL
For Use With (Application) Augmenting Cell Culture Growth
Heat Inactivated Yes
Hemoglobin Concentration ≤ 25 mg/dL
Product Line HyClone
Shipping Condition Frozen
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.