Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin

Catalog No. SH304010240 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Processing:
Heat inactivated
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH304010240 1000 mL Irradiated, 25 to 40 doses
SH304010140 500 mL Irradiated, 25 to 40 doses
SH3040102HI 1000 mL Heat inactivated
SH3040101HI 500 mL Heat inactivated
SH304010245 1000 mL Irradiated, 30 to 45 doses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SH304010240 Supplier Cytiva Supplier No. SH30401.02IR2540
Only null left
Add to Cart
Add to Cart

A cost-effective alternative to Fetal Bovine Serum (FBS).

  • Carefully collected, processed, and filtered in New Zealand
  • Ensured safety against bovine diseases
  • We follow the same standards in processing of our calf serum as with our fetal bovine serum (FBS) to enable offering the highest quality and traceability
  • HyClone Newborn Calf Serum, New Zealand Origin is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters.
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Additional Assays Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Iron Saturation,Glucose,IgG
    Certifications/Compliance USDA Regulations
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
    Quantity 1000 mL
    Sterility Sterile
    Sterilization Method 0.1 μm filtration
    Processing Irradiated, 25 to 40 doses
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 25 EU/mL
    Heat Inactivated No
    Mycoplasma Detected Yes
    USDA Safety Testing Included Yes
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.