Learn More
Description
Specifications
Specifications
| Additional Assays | Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Iron Saturation,Glucose,IgG |
| Certifications/Compliance | USDA Regulations |
| Color | Amber |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C |
| Species | Bovine |
| Age | Newborn (20 days or less) |
| Country of Origin | New Zealand |
| Format | Bottle |
| Product Type | Newborn Calf Bovine Serum |
| Quantity | 1000 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.