Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Newborn Calf Serum (U.S.)

Catalog No. SH3011802 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Irradiated, 25 to 40 doses
None
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3011802 100 mL None
SH3011804 1000 mL None
SH3011803 500 mL None
SH301180440 1000 mL Irradiated, 25 to 40 doses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. SH3011802 Supplier Cytiva Supplier No. SH30118.02
Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Augment cell culture growth with U.S.-sourced HyClone™ Newborn Bovine Calf Serum.

  • Filtered through sequential 100nm filters and sourced from animals that are typically less than 10 days old
TRUSTED_SUSTAINABILITY

Specifications

Description Newborn Calf Serum (U.S.)
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.