Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ SFM4MegaVir Powdered Media

Catalog No. SH3058701 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3058701 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3058701 Supplier Cytiva Supplier No. SH30587.01

Increase process yields in the manufacture of viral vaccines

HyClone SFM4MegaVir is a regulatory-friendly and animal-derived component-free (ADCF) cell culture medium. The medium was designed to increase process yields in the manufacture of viral vaccines in a variety of anchorage dependent cell lines including Vero, MDCK, COS-7, and MDBK cells.

SFM4MegaVir has been successfully tested in a variety of culture systems for cell growth and virus production including T-flasks and suspension microcarrier cultures.

SFM4MegaVir is available in liquid and powder formats in user-friendly packaging.

  • ADCF cell culture medium
  • Designed for high viral vaccine production
  • Suitable for direct adaptation
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Sterility Non-sterile
With Additives Phenol Red
Without Additives No Glutamine, No Sodium Bicarbonate
Quantity 5 L
Content And Storage 2 to 8°C, away from light and moisture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.