Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Water for Injection (WFI) Quality Water

Catalog No. SH3022117 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
20 L
50 L
6 x 1000 mL
100 L
200 L
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3022117 500 mL
SH3022124 10 L
SH3022125 20 L
SH3022126 50 L
SH3022127 100 L
SH3022128 200 L
SH3022110 1 L
SH30221LS 6 x 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. SH3022117 Supplier Cytiva Supplier No. SH30221.17
This item is not returnable. View return policy

Supports cell culture growth and biomanufacturing process.

  • Format: Liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
SMILES O

Specifications

IUPAC Name water
Molecular Weight (g/mol) 18.02
Quantity 500 mL
Chemical Name or Material Water
ShelfLife 36 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.