Learn More
Hydra-Point™ Coulometric Generator Solution, Universal, BAKER ANALYZED™ Reagent, J.T. Baker™
Shop All Avantor ProductsDescription
- Catholyte solution for coulometric analysis. For both general use and aldehydes and ketones.
Specifications
Specifications
| Flash Point | 46.00°C |
| Packaging | Glass Ampoule |
| Quantity | 5 mL |
| Grade | BAKER ANALYZED™ Reagent |
| Chemical Name or Material | Hydra-Point™ Coulometric Generator Solution, Universal |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.