Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydra-Point™ Coulometric Generator Solution, Universal, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003097 Shop All Avantor Products
Change view
Click to view available options
Quantity:
5 mL
125 mL
Packaging:
Glass Ampoule
Narrow Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-097 125 mL Narrow Mouth Amber Glass Bottle
02-003-094 5 mL Glass Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-003-097 Supplier Avantor Supplier No. 628600

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

High quality chemicals for laboratory and specialized industrial use

    Catholyte solution for coulometric analysis. For both general use and aldehydes and ketones.

Specifications

Flash Point 46.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 125 mL
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Hydra-Point™ Coulometric Generator Solution, Universal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.