Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydralazine Hydrochloride, 99%, Spectrum™ Chemical
SDP

Catalog No. 18603146 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-146 25 g Amber Glass Bottle
18-603-145 5 g Amber Glass Bottle
18-603-147 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-146 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HO12025GM
Add to Cart
Add to Cart

HO120, 304-20-1, C8H8N4·HCl

Spectrum™ Chemical Hydralazine Hydrochloride has been used to treat hypertension and can act as a vasodilator. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 304-20-1
Molecular Formula C8H9ClN4
Molecular Weight (g/mol) 196.64
InChI Key ZUXNZUWOTSUBMN-UHFFFAOYSA-N
IUPAC Name hydrogen 1-hydrazinylphthalazine chloride
SMILES [H+].[Cl-].NNC1=C2C=CC=CC2=CN=N1

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
UN Number UN2811
Percent Purity 99%
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Hydralazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.