Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydralazine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18603150 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-150 100 g Amber Glass Bottle
18-603-148 5 g Amber Glass Bottle
18-603-149 25 g Amber Glass Bottle
18-603-151 1 kg Amber Glass Bottle
18-603-152 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-150 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H1289100GM
Add to Cart
Add to Cart

H1289, 304-20-1, C8H8N4·HCl

Spectrum™ Chemical Hydralazine Hydrochloride, USP has been used to treat hypertension because it can act as a vasodilator mostly affecting arterioles and arteries. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 304-20-1
Molecular Formula C8H9ClN4
Molecular Weight (g/mol) 196.64
InChI Key ZUXNZUWOTSUBMN-UHFFFAOYSA-N
IUPAC Name hydrogen 1-hydrazinylphthalazine chloride
SMILES [H+].[Cl-].NNC1=C2C=CC=CC2=CN=N1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 100 g
UN Number UN2811
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.