Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™-Solvent E, Solstice

Catalog No. 6000668 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-006-68 500 mL
60-006-66 1 L
60-006-67 2.5 L
60-046-402 6 x 500 mL
60-046-401 6 x 1 L
60-046-400 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-006-68 Supplier Solstice Supplier No. 34730500ML
Add to Cart
Add to Cart

Medium for volumetric two-component Karl Fischer titration (ethanol-based)

Advantages of Hydranal two-component reagents:

  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents: reduced toxicity compared to methanol
  • Long shelf life (three years for titrants, five years for solvents)

Improved Composition

In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Solvent E
CAS 288-32-4, 111-42-2, 64-17-5, 7446-09-5
Density 0.90g/mL
Quantity 500 mL
Flash Point 12°C (53.6°F)
MDL Number MFCD01867183
UN Number UN1170
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.