Learn More
HYDRANAL™ - Solvent Oil, Reagent for volumetric two-component KF titration in oils (working medium), Solstice
Shop All Honeywell Research Chemicals ProductsDescription
Advantages of Hydranal two-component reagents:
- High titration speed
- Ideal accuracy for small amounts of water
- High buffer capacity
- Exact and stable titer
- E-type reagents: reduced toxicity compared to methanol
- Long shelf life (three years for titrants, five years for solvents)
Improved Composition
In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™ - Solvent Oil |
| Name Note | Reagent for volumetric two-component KF titration in oils (working medium) |
| Boiling Point | 63°C |
| CAS Min % | ≥1.0000%, ≥30.0000% |
| CAS Max % | <10.0000%, <5.0000% |
| CAS | 288-32-4, 67-56-1, 111-27-3, 7446-09-5 |
| Density | ∼0.83 g/mL |
| Quantity | 6 x 1 L |
| Flash Point | 17°C |
| Packaging | Glass Bottle |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.