Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Hydrated-Sponge, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. S26778 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Includes:
Gloves HS10BPW2G
Gloves HS10LET2G
Gloves HS10NB2G
Saturation Solution:
10 mL Buffered Peptone Water Broth
10 mL letheen broth
10 mL neutralizing buffer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Saturation Solution
S26778 Gloves HS10NB2G 10 mL neutralizing buffer
S26774 Gloves HS10BPW2G 10 mL Buffered Peptone Water Broth
S26776 Gloves HS10LET2G 10 mL letheen broth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S26778 Supplier 3M™ Supplier No. HS10NB2G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uses simplified packaging to reduce waste and minimize handling and preparation

  • Ready-to-use, pre-hydrated sponges for easy and convenient environmental and food sample collection
  • Durable sponge withstands scrubbing
  • Sponge bag with write-on label for ease in labeling samples
  • Includes two decontaminated gloves for safety and convenience

Specifications

Product Type Hydrated Sponge
Length (English) 3 in.
Length (Metric) 7.6 cm
Width (English) 1.5 in.
Width (Metric) 4 cm
Packaging Quantity 100/Cs.
Saturation Solution 10 mL neutralizing buffer
Includes Gloves HS10NB2G
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.