Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine Acetate 98.0+%, TCI America™
SDP

Catalog No. H111225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H111225G 25 g
H11125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H111225G Supplier TCI America Supplier No. H111225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13255-48-6
Molecular Formula C2H8N2O2
Molecular Weight (g/mol) 92.10
MDL Number MFCD00013141
InChI Key YFHNDHXQDJQEEE-UHFFFAOYSA-N
PubChem CID 165591
IUPAC Name acetic acid; hydrazine
SMILES NN.CC(O)=O

Specifications

Melting Point 101°C
Color White
Quantity 25 g
UN Number 3263
Formula Weight 92.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hydrazine Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.