Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine dihydrochloride, 98+%

Catalog No. AA3272522
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3272522 100 g
AA3272536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3272522 Supplier Thermo Scientific Chemicals Supplier No. 03272522
Add to Cart

missing translation for 'validMainframeMsg'

For Use As Chlorine Scavenger in Hcl Gas Streams | CAS: 5341-61-7 | ClH5N2 | 68.50 g/mol

Hydrazine dihydrochloride is used as chlorine scavenger in HCl gas streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5341-61-7
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.50
MDL Number MFCD00064543
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
Synonym hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
PubChem CID 17548
IUPAC Name hydrazine;dihydrochloride
SMILES [H+].[Cl-].NN

Specifications

Boiling Point 200°C (decomposition)
Melting Point 198°C
Color White
Physical Form Lump
Quantity 100 g
Assay Percent Range >98%
Solubility Information Soluble in water. Slightly soluble in alcohol
Formula Weight 104.97
Odor Odorless
Appearance White crystalline or lump
Density 1.42g/cm3
Chemical Name or Material Hydrazine dihydrochloride, Special handling precautions required
Show More Show Less
Hazard Category H301+H311+H331-H317-H350
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
DOTInformation DOT Class: 6.1, Packing Group: III
EINECSNumber 226-283-2
RTECSNumber MV2298000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.