Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. H017025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H017025G 25 g
H0170500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H017025G Supplier TCI America Supplier No. H017025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5341-61-7
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.50
MDL Number MFCD00064543
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
Synonym hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
PubChem CID 17548
IUPAC Name hydrogen hydrazine chloride
SMILES [H+].[Cl-].NN

Specifications

Physical Form Powder/Crystals
UN Number 3290
Color White
Melting Point 200°C
Quantity 25 g
Percent Purity ≥98.0% (T)
Chemical Name or Material Hydrazine Dihydrochloride
EINECSNumber (1)-0374
RTECSNumber MV2298000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.