Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine monohydrate, 99+%

Catalog No. AA1665157
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
4 x 500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1665157 4 x 500 g
AA1665122 100 g
AA1665136 500 g
AA16651A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1665157 Supplier Thermo Scientific Chemicals Supplier No. 01665157
Add to Cart

CAS: 7803-57-8 | H6N2O | 50.06 g/mol

Hydrazine Hydrate used as a reactant in the cyclizations of pyridinones. It is also used in the study of nanocrystal semiconductors, participating in the functionalization and passivation of surface states. It is widely used as a reducing agent or an intermediate of synthesis in various industrial sectors like water treatment (effluents, industrial boilers), chemical treatment process (metals, mine extraction) or active ingredients synthesis (pharmaceuticals and agrochemicals).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-57-8
Molecular Formula H6N2O
Molecular Weight (g/mol) 50.06
MDL Number MFCD00149931
InChI Key IKDUDTNKRLTJSI-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
SMILES O.NN

Specifications

Boiling Point 120°C to 121°C
Melting Point -52°C
Color Colorless
Physical Form Liquid
Quantity 4 x 500 g
Assay Percent Range >99%
Linear Formula H2NNH2·H2O
UN Number UN2030
Flash Point 75° (167°F)
Merck Index 14,4771
Solubility Information Soluble in chloroform,DMSO,methanol. Slightly soluble in water.
IUPAC Name hydrazine;hydrate
Formula Weight 50.06 (32.05 Anhydrous)
Percent Purity ≥99%
Odor Amine-like
Refractive Index 1.4310
Density 1.027g/mL
Chemical Name or Material Hydrazine monohydrate
Show More Show Less
Hazard Category H227-H301+H311-H314-H317-H330-H335-H350
Hazard Statement GHS H Statement
H301-H310-H330-H350-H314-H318-H227-H317 Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280g-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDRAZINE AQUEOUS SOLUTION
EINECSNumber 206-114-9
RTECSNumber MV4590000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.