Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine Sulfate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18603154 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
6 x 125 g
6 x 500 g
12 kg
50 kg
Packaging:
Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-154 25 g Poly Bottle
18-603-155 125 g Poly Bottle
18-603-157 500 g Poly Bottle
18-603-156 6 x 125 g Poly Bottle
18-603-158 6 x 500 g Poly Bottle
18-603-159 12 kg Poly Pail
18-603-160 50 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-603-154 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H101825GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

H1018, 10034-93-2, H4N2·H2SO4

Spectrum™ Chemical Hydrazine Sulfate, Crystal, Reagent, ACS has been promoted to treat colon and rectal (colorectal) cancer, lung cancer, brain cancer (neuroblastoma), Hodgkin′s disease (lymph cancer), and other cancers. It is also used for the general weight loss and wasting (cachexia) associated with cancer. However, hydrazine sulfate is not approved for use with cancer patients in the United States. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 10034-93-2
Molecular Formula H6N2O4S
Molecular Weight (g/mol) 130.12
InChI Key ZGCHATBSUIJLRL-UHFFFAOYSA-N
IUPAC Name hydrazine; sulfuric acid
SMILES NN.OS(O)(=O)=O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 25 g
UN Number UN1759
Percent Purity 99%
Grade ACS
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.