Learn More
Sigma Organic Chemistry hydrazino-acetic ethyl | 50mg | 637-80-9 | MFCD00044588
Supplier: Sigma Organic Chemistry CDS00991250MG

hydrazino-acetic ethyl , 50mg
About this Item:
CAS #: 637-80-9
MDL #: MFCD00044588
Molecular Weight: 118.13
UNSPSC Code: 12352200
Chemical Formula: C4H10N2O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.