Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrindantin Hydrate, Approx. 96%, Spectrum™ Chemical
SDP

Catalog No. 18603162 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-162 25 g Amber Glass Bottle
18-603-161 5 g Amber Glass Bottle
18-603-163 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-162 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H245125GM
Add to Cart
Add to Cart

H2451, 5950-69-6, C18H10O6·2H2O

Spectrum™ Chemical Hydrindantin Hydrate

Chemical Identifiers

CAS 5950-69-6
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
InChI Key QHVADKNWNMILPQ-UHFFFAOYNA-N
IUPAC Name 2,2',3,3,3',3'-hexahydroxy-1H,1'H,2H,2'H,3H,3'H-[2,2'-biindene]-1,1'-dione
SMILES OC1(O)C2=CC=CC=C2C(=O)C1(O)C1(O)C(=O)C2=CC=CC=C2C1(O)O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity ∼96%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.