Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrobromic Acid, 47.0%, Suprapur™, MilliporeSigma™

Catalog No. M1003060250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1003060250 250 mL
M1003061000 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1003060250 Supplier MilliporeSigma Supplier No. 1.00306.0250
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym Hydrogen bromide solution
IUPAC Name hydrogen bromide
SMILES Br

Specifications

Melting Point −11°C
Boiling Point 126°C (1013 hPa)
Density 1.49 g/cm3 (at 20°C)
Packaging Glass Bottle
Vapor Pressure 10.6 hPa (20°C)
Assay Percent Range ≥ 47%
Quantity 250 mL
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.