Learn More
Description
- For Laboratory,Research,or Manufacturing Use
Specifications
Specifications
| Packaging | Narrow Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 1.50kg |
| Quantity | 4 L |
| IUPAC Name | bromane |
| Formula Weight | 80.92 |
| Percent Purity | 47 to 49% |
| Grade | Meets ACS reagent chemical requirements |
| Chemical Name or Material | Hydrobromic Acid |
Safety and Handling
| Recommended Storage | Material darkens when exposed to air and light |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.