Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrobromic Acid, 47-49% (Certified ACS), Fisher Chemical™

Catalog No. A140100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A140-100 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A140-100

missing translation for 'validMainframeMsg'

Note: This reagent may darken on standing

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6 , 7732-18-5
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name hydrogen bromide
SMILES Br

Specifications

pH <1
Color Yellow
Melting Point -11°C
Boiling Point 126°C
Density 1.480g/cm³
Heavy Metals (as Pb) 5ppm max.
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 8 mmHg at 25°C
Assay Percent Range 47.0 to 49.0 %
Quantity 100 mL
Physical Form Liquid
UN Number 1788
Formula Weight 80.91
Percent Purity 47.0 to 49.0%
Grade Certified ACS
Chemical Name or Material Hydrobromic Acid, 47-49%
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.