Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.005 N (N/200) in Methanol, Ricca Chemical
SDP

Catalog No. R38095004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R38095004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R38095004A Supplier Ricca Chemical Company Supplier No. R38095004A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standardized hydrochloric acid solution in methanol for general laboratory reagent applications.

Chemical Identifiers

CAS 67-56-1 , 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless
CAS Min % 0.06, 97.0, 0.57
CAS Max % 0.06, 100.0, 0.6
Packaging Natural Poly Bottle
Quantity 4 L
Physical Form Liquid
Grade Not Available
Concentration or Composition (by Analyte or Components) 0.005 Normal (N/200) in Methanol
Chemical Name or Material Hydrochloric Acid
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.