Learn More
Specifications
Specifications
| Color | Colorless |
| Odor | Odorless to slightly pungent |
| Packaging | Cubitainer |
| Solvent or Matrix | Aqueous Solution |
| Assay Percent Range | 0.01000 ± 0.00005 N at 20°C |
| Quantity | 4 L |
| Physical Form | Liquid |
| Solubility Information | Solubility in Water: Infinite |
| Grade | Laboratory |
| Chemical Name or Material | Hydrochloric Acid, 0.0100 Normal |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
