Learn More
Hydrochloric acid, 0.01N Standardized Solution
Description
Hydrochloric acid, 0.01N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Physical Form | Liquid |
| Color | Colorless |
| Solubility | Miscible with water, methanol,ethanol and ether |
| IUPAC Name | hydrogen chloride |
| Quantity | 4 L |
| Formula Weight | 36.46 |
| Concentration or Composition (by Analyte or Components) | Hydrochloric acid: 0.083%; Water: 99.917% |
| Chemical Name or Material | Hydrochloric acid, 0.01N |
Safety and Handling
| EINECSNumber | 231-595-7 |
| RTECSNumber | MW4025000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.