Learn More
Hydrochloric Acid, 0.0200 N (N/50) in Isopropyl Alcohol, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Color | Colorless |
| Concentration | 0.0200Normal |
| CAS Min % | 0.22, 96.0, 2.27 |
| CAS Max % | 0.23, 99.0, 2.36 |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Isopropyl Alcohol |
| Quantity | 4 L |
| Physical Form | Liquid |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid |
| Grade | Laboratory |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.