Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 0.025 N, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88604 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S88604 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88604 Supplier Aqua Phoenix Scientific Supplier No. HA6025P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standardized hydrochloric acid solution for laboratory analysis.

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Concentration 0.025 N
Packaging Bottle
Quantity 500 mL
Grade ACS
Chemical Name or Material Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.