Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid, 0.5N Standardized Solution

Catalog No. AA35642M4
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35642M4 20 L
AA35642K2 1 L
AA35642K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35642M4 Supplier Thermo Scientific Chemicals Supplier No. 035642M4
Add to Cart

CAS: 7647-01-0 | ClH | 36.46 g/mol

Hydrochloric acid, 0.5N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl

Specifications

Appearance Colorless liquid
Color Colorless
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Quantity 20 L
Physical Form Liquid
Solubility Information Miscible with water at room temperature. Also miscible with methanol,ethanol and ether.
Formula Weight 36.46
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 4.13%; Water: 95.87%
Chemical Name or Material Hydrochloric acid
Hazard Category H290-H315-H318
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 231-595-7
RTECSNumber MW4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.