Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 1.0 N Aqueous Solution, APHA, 1 N +/-0.005 N, Spectrum™ Chemical
SDP

Catalog No. 18603170 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-170 4 x 4 L Poly Bottle
18-603-167 1 L Poly Bottle
18-603-168 4 L Poly Bottle
18-603-169 20 L Poly Bottle
18-603-171 200 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-170 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H1354LTPLCS4
Add to Cart
Add to Cart

H-135, 7647-01-0

Spectrum™ Chemical Hydrochloric Acid, 1.0 N Aqueous Solution, APHA , is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids it is the least likely to be subject to an oxidation-reduction reaction. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis.

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Concentration 1 N ±0.005
CAS Max % 3.6 to 3.7%, 96.3 to 96.4%
Packaging Poly Bottle
Quantity 4 x 4 L
UN Number UN1789
Grade Ungraded
Chemical Name or Material Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.