Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 12.0 N, Ricca Chemical
SDP

Catalog No. 37805PT Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Natural Poly Bottle
Safety-Coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
37805PT 2.5 L Safety-Coated Glass Bottle
R37800001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37805PT Supplier Ricca Chemical Company Supplier No. 37805PT
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883

Specifications

Color Colorless to Green-Yellow
Concentration 12.0Normal
CAS Min % 98.0
CAS Max % 100.0
Packaging Safety-Coated Glass Bottle
Quantity 2.5 L
Molecular Formula ClH
Physical Form Liquid
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
Grade Laboratory
Chemical Name or Material Hydrochloric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.