Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 15% w/w, Reagents

Catalog No. 01335009
Change view
Click to view available options
Quantity:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-009 20 L
01-335-010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-009 Supplier Reagents Holdings Llc Supplier No. CH12910020F
Add to Cart
Add to Cart

Hydrochloric Acid, 15% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH 0
Color Colorless to Slight Yellow
Packaging Cubitainer
Specific Gravity 1.0745
Quantity 20 L
Physical Form Liquid
Percent Purity 15%
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 15% w/w
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.