Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 4% v/v in Methanol, Reagents

Catalog No. 01335145
Change view
Click to view available options
:
Quantity: 2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-145 Quantity: 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-145 Supplier Reagents Holdings Llc Supplier No. CH0104002.5D1
Only null left
Add to Cart
Add to Cart

Hydrochloric Acid, 4% v/v in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name methanol
SMILES CO

Specifications

pH <1
Color Colorless
Boiling Point 65°C
Packaging Glass Bottle
Specific Gravity 0.81
Quantity 2.5 L
Physical Form Liquid
Percent Purity 4%
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 4% v/v in Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.