Learn More
Specifications
Specifications
| Color | Colorless to Green-Yellow |
| Appearance of Solution | Colorless to slightly Greenish-yellow Liquid |
| Concentration | 4.995 to 5.005 N |
| CAS Min % | 66.0, 32.21 |
| CAS Max % | 68.0, 33.53 |
| Packaging | Natural Poly Bottle |
| Quantity | 4 L |
| Physical Form | Liquid |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| Molecular Weight (g/mol) | 36.46 |
| Show More |
Safety and Handling
| ShelfLife | 18 months |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
