Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 5.00 N, Ricca Chemical
SDP

Catalog No. R374000010F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Natural Poly Bottle
Safety-coated Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R374000010F 10 L Cubitainer
R3740000500 500 mL Natural Poly Bottle
374032G 1 L Amber Glass Bottle
3740-32 1 L Natural Poly Bottle
37405PT 2.5 L Safety-coated Glass Bottle
R37400004F 4 L Cubitainer
3740-1 4 L Natural Poly Bottle
3740-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. R374000010F Supplier Ricca Chemical Company Supplier No. R374000010F
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless to Green-Yellow
Appearance of Solution Colorless to slightly Greenish-yellow Liquid
Concentration 4.995 to 5.005 N
CAS Min % 66.0, 32.21
CAS Max % 68.0, 33.53
Packaging Cubitainer
Quantity 10 L
Synonym hydrochloric acid
Molecular Weight (g/mol) 36.46
Grade Laboratory
Assay Normal at 20°C
Chemical Name or Material Hydrochloric Acid
Show More Show Less
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.