Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 5% (v/v) in Denatured Alcohol, Ricca Chemical
SDP

Catalog No. 356815 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Cubitainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
356815 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 356815 Supplier Ricca Chemical Company Supplier No. 3568.15
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 67-56-1 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
PubChem CID 313
ChEBI CHEBI:23116
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless
CAS Min % 7.06, 91.0
CAS Max % 7.35, 95.0
Packaging Cubitainer
Quantity 20 L
Physical Form Liquid
Percent Purity 5% V/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% (v/v) in Denatured Alcohol (1 + 19)
Chemical Name or Material Hydrochloric Acid
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.