Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, 6.25 N, Ricca Chemical
SDP

Catalog No. 37555 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Natural Poly Bottle
Safety-coated Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3755-5 20 L Cubitainer
R3755000500 500 mL Natural Poly Bottle
375532G 1 L Amber Glass Bottle
3755-32 1 L Natural Poly Bottle
37555PT 2.5 L Safety-coated Glass Bottle
37551G 4 L Amber Glass Bottle
3755-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 3755-5 Supplier Ricca Chemical Company Supplier No. 37555
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless to Green-Yellow
Concentration 6.25Normal
CAS Min % 61.0, 37.21
CAS Max % 63.0, 38.73
Packaging Cubitainer
Quantity 20 L
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Hydrochloric Acidl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.