Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, ACS Reagent, 37%, Solstice

Catalog No. 6000705 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
4 x 4 L
Density:
1.2 g/mL at 25°C
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Density
60-007-05 2.5 L 1.2 g/mL at 25°C
60-007-10 500 mL 1.2 g/mL at 25°C
60-007-12 6 x 500 mL 1.2 g/mL at 25°C
60-007-09 4 x 4 L 1.2 g/mL at 25°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-007-05 Supplier Solstice Supplier No. 2581482.5L
Add to Cart
Add to Cart

ACS Reagent, 37%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Concentration 37%
Density 1.2 g/mL at 25°C
Residue after Ignition ≤5ppm
Vapor Pressure 3.23psi (21.1°C), 7.93psi (37.7°C)
Quantity 2.5 L
UN Number 1789
Beilstein 1098214
IUPAC Name hydrogen chloride
Formula Weight 36.46g/mol
Grade ACS Reagent
Chemical Name or Material Hydrochloric Acid
Show More Show Less
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H290-H314-H335
EINECSNumber 231-595-7
RTECSNumber MW4025000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.