Learn More
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Concentration | 37% |
| Density | 1.2 g/mL at 25°C |
| Residue after Ignition | ≤5ppm |
| Vapor Pressure | 3.23psi (21.1°C), 7.93psi (37.7°C) |
| Quantity | 6 x 500 mL |
| UN Number | 1789 |
| Beilstein | 1098214 |
| IUPAC Name | chlorane |
| Formula Weight | 36.46g/mol |
| Grade | ACS Reagent |
| Show More |
Safety and Handling
| Hazard Statement | P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 |
| Precautionary Statement | H290-H314-H335 |
| EINECSNumber | 231-595-7 |
| RTECSNumber | MW4025000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.