Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid solution, Volumetric, Reag. Ph. Eur., 1 M HCl (1N), Solstice

Catalog No. 6000747 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
5 L
10 L
30 L
4 x 5 L
Packaging:
Plastic Bottle
Plastic Pail
VOLPAC™ CONTAINER
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-007-47 10 L VOLPAC™ CONTAINER
60-007-48 1 L Plastic Bottle
60-007-52 5 L VOLPAC™ CONTAINER
60-007-51 5 L Plastic Bottle
60-046-562 4 x 5 L Plastic Bottle
60-007-50 30 L Plastic Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-007-47 Supplier Solstice Supplier No. 3532810LVP
Add to Cart
Add to Cart

Volumetric, Reag. Ph. Eur., 1 M HCl (1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Concentration 1M
Packaging VOLPAC™ CONTAINER
Vapor Pressure 190 mmHg (25°C)
Quantity 10 L
UN Number UN1789
IUPAC Name chlorane
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric acid solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.