Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochlorothiazide, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18603219 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-219 12 kg Amber Glass Bottle
18-603-216 25 g Amber Glass Bottle
18-603-217 100 g Amber Glass Bottle
18-603-218 500 g Amber Glass Bottle
18-603-220 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-219 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HY10812KGBL
Add to Cart
Add to Cart
Request a Quote

HY108, 58-93-5, C7H8ClN3O4S2

Spectrum™ Chemical Hydrochlorothiazide, USP is part of the thiazide class of diuretic drugs used for the treatment of congestive heart failure and hypertension. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-93-5
Molecular Formula C7H8ClN3O4S2
Molecular Weight (g/mol) 297.73
MDL Number MFCD00051765
InChI Key JZUFKLXOESDKRF-UHFFFAOYSA-N
IUPAC Name 6-chloro-1,1-dioxo-3,4-dihydro-2H-1λ6,2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)C1=CC2=C(NCNS2(=O)=O)C=C1Cl

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 12 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Hydrochlorothiazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.