Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrocortisone 17-Butyrate 98.0+%, TCI America™
SDP

Catalog No. H15061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15061G 1 g
H1506200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15061G Supplier TCI America Supplier No. H15061G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13609-67-1
Molecular Formula C25H36O6
Molecular Weight (g/mol) 432.557
MDL Number MFCD00083364
InChI Key BMCQMVFGOVHVNG-TUFAYURCSA-N
Synonym 11beta,17,21-Trihydroxypregn-4-ene-3,20-dione 17-Butyrate, Cortisol 17-Butyrate
PubChem CID 26133
ChEBI CHEBI:31674
IUPAC Name [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] butanoate
SMILES CCCC(=O)OC1(CCC2C1(CC(C3C2CCC4=CC(=O)CCC34C)O)C)C(=O)CO

Specifications

Melting Point 212°C
Color White
Quantity 1 g
Formula Weight 432.56
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Hydrocortisone 17-Butyrate
RTECSNumber GM9002000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.