Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Getinge USA HYDROCYDE H2O2 250ML

Catalog No. NC1805719 Shop All Getinge USA Products
Encompass

  • Effective sterilizing agent for rigorous decontamination.
  • Designed for industrial sterilization applications.
  • Contains 35% hydrogen peroxide as the active ingredient.
  • Aqueous solution specifically formulated for scientific and medical environments.
  • Supplied in a volume of 250 mL.
  • Typically supplied as a pack of 10 units.
  • Chemical formula is H2O2.
  • Classified as a substance chemical type.
  • Ensures high-quality sterilization performance.

Catalog No. NC1805719 Supplier Getinge USA Supplier No. 6007012118
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Effective sterilizing agent for rigorous decontamination.
  • Designed for industrial sterilization applications.
  • Contains 35% hydrogen peroxide as the active ingredient.
  • Aqueous solution specifically formulated for scientific and medical environments.
  • Supplied in a volume of 250 mL.
  • Typically supplied as a pack of 10 units.
  • Chemical formula is H2O2.
  • Classified as a substance chemical type.
  • Ensures high-quality sterilization performance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.