Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrofluoric Acid, 2% (v/v), Ricca Chemical
SDP

Catalog No. 3819232 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3819-232 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3819-232 Supplier Ricca Chemical Company Supplier No. 3819.232
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES F

Specifications

Color Colorless
CAS Min % 96.0, 2.22
CAS Max % 100.0, 2.31
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Percent Purity 2% V/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 2% (v/v) Aqueous Solution
Chemical Name or Material Hydrofluoric Acid
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.